6-Brom-m-anisaldehyd 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-methoxybenzaldehyde

OR5449-1G
APOSOR5449-1GEA 21.9 EUR
APOSOR5449-1G
6-Brom-m-anisaldehyd 97%
6-Brom-m-anisaldehyd
Formel: C₈H₇BrO₂
Molecular Weight: 215.05 g/mol
Schmelzpunkt: 71…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00463817
CAS-Nummer: 7507-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR