5-Iodcytosin
Lieferant: Apollo Scientific
|
Achtung
|
OR54491-5G
APOSOR54491-5GEA
68.3
EUR
APOSOR54491-5G
5-Iodcytosin
5-Iodcytosin
Formel:
C₄H₄IN₃O Schmelzpunkt: 250 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00023162 CAS-Nummer: 1122-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...