2,4'-Dichlorbenzophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4'-DBP

OR54515-500G
APOSOR54515-500GEA 213 EUR
APOSOR54515-500G
2,4'-Dichlorbenzophenon 98%
2,4'-Dichlorbenzophenon
Formel: C₁₃H₈Cl₂O
Molecular Weight: 251,11 g/mol
Schmelzpunkt: 63…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038744
CAS-Nummer: 85-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR