5'-Hydroxy-2'-methoxyacetophenon

Lieferant: Apollo Scientific

Synonyme: 1-(5-Hydroxy-2-methoxyphenyl)ethanone

OR54528-1G OR54528-250MG OR54528-5G
APOSOR54528-1GEA 87.6 EUR
APOSOR54528-1G APOSOR54528-250MG APOSOR54528-5G
5'-Hydroxy-2'-methoxyacetophenon
5'-Hydroxy-2'-methoxyacetophenon
Formel: C₉H₁₀O₃
Molecular Weight: 166.1739 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038299
CAS-Nummer: 31405-60-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR