tert-Butyl 3-(4-aminophenyl)piperidine-1-carboxylate 95%

Lieferant: Apollo Scientific

OR54550-1G
APOSOR54550-1GEA 502 EUR
APOSOR54550-1G
tert-Butyl 3-(4-aminophenyl)piperidine-1-carboxylate 95%
tert-Butyl 3-(4-aminophenyl)piperidine-1-carboxylate
Formel: C₁₆H₂₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05861561
CAS-Nummer: 875798-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR