2,3-Dihydroxy-4-methoxybenzoic acid

Lieferant: Apollo Scientific

OR54556-1G
APOSOR54556-1GEA 441 EUR
APOSOR54556-1G
2,3-Dihydroxy-4-methoxybenzoic acid
2,3-Dihydroxy-4-methoxybenzoic acid
Formel: C₈H₈O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00016499
CAS-Nummer: 3934-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR