O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine 98%

Lieferant: Apollo Scientific

Synonyme: N-Fmoc-O-tert-butyl-L-serine, N-a-Fmoc-O-t-butyl-L-serine, Fmoc-Ser(tBu)-OH, O-tert-Butyl-N-Fmoc-L-serine

OR54566-100G
APOSOR54566-100GEA 85.7 EUR
APOSOR54566-100G
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine 98%
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine
Formel: C₂₂H₂₅NO₅
Molecular Weight: 383,44 g/mol
Schmelzpunkt: 127…136 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037127
CAS-Nummer: 71989-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR