N-Fmoc-glycin 98%

Lieferant: Apollo Scientific

Synonyme: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH

OR54569-500G
APOSOR54569-500GEA 169 EUR
APOSOR54569-500G
N-Fmoc-glycin 98%
N-Fmoc-glycin
Formel: C₁₇H₁₅NO₄
Molecular Weight: 297.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037140
CAS-Nummer: 29022-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR