Fmoc-L-phenylalanin 98%

Lieferant: Apollo Scientific

Synonyme: N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, N-α-Fmoc-L-Phenylalanine, Fmoc-Phe-OH, N-Fmoc-L-phenylalanine

OR54571-500G
APOSOR54571-500GEA 219 EUR
APOSOR54571-500G
Fmoc-L-phenylalanin 98%
Fmoc-L-phenylalanin
Formel: C₂₄H₂₁NO₄
Molecular Weight: 387.4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037128
CAS-Nummer: 35661-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR