Nα-Fmoc-N-δ-trityl-L-glutamine 98%

Lieferant: Apollo Scientific

Synonyme: Fmoc-Gln(Trt)-OH

OR54572-100G
APOSOR54572-100GEA 105 EUR
APOSOR54572-100G
Nα-Fmoc-N-δ-trityl-L-glutamine 98%
N-alpha-Fmoc-N-delta-trityl-L-glutamine
Formel: C₃₉H₃₄N₂O₅
Molecular Weight: 610,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077056
CAS-Nummer: 132327-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR