Fmoc-O-tert-butyl-D-serine 97%

Lieferant: Apollo Scientific

Synonyme: N-Fmoc-O-tert-butyl-D-serine, Fmoc-D-Ser(tBu)-OH

OR54574-25G
APOSOR54574-25GEA 81.2 EUR
APOSOR54574-25G
Fmoc-O-tert-butyl-D-serine 97%
Fmoc-O-tert-butyl-D-serine
Formel: C₂₂H₂₅NO₅
Molecular Weight: 383,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077071
CAS-Nummer: 128107-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR