Methyl-1-benzyl-4-oxo-3-piperidincarboxylathydrochlorid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, 1-Benzyl-4-oxo-3-piperidinecarboxylic acid methyll ester hydrochloride, Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate HCl, Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride

OR54576-100G
APOSOR54576-100GEA 105 EUR
APOSOR54576-100G
Methyl-1-benzyl-4-oxo-3-piperidincarboxylathydrochlorid 95+%
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Formel: C₁₄H₁₈ClNO₃ C₁₅H₁₉NO₃·HCl
Molecular Weight: 283,75 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012799
CAS-Nummer: 3939-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR