3-Ethynylimidazo[1,2-b]pyridazine
Lieferant: Apollo Scientific
|
Achtung
|
OR54580-5G
APOSOR54580-5GEA
119
EUR
APOSOR54580-5G
3-Ethynylimidazo[1,2-b]pyridazine
3-Ethynylimidazo[1,2-b]pyridazine
Intermediate in Ponatinib synthesis.
Formel:
C₈H₅N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD17012027 CAS-Nummer: 943320-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...