Ethyl (6-aminopyridin-2-yl)acetate

Lieferant: Apollo Scientific

OR54597-1G
APOSOR54597-1GEA 364 EUR
APOSOR54597-1G
Ethyl (6-aminopyridin-2-yl)acetate
Ethyl (6-aminopyridin-2-yl)acetate
Formel: C₉H₁₂N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD09757476
CAS-Nummer: 71469-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR