3-Acetylthiazolidine-2-thione
Lieferant: Apollo Scientific
|
Achtung
|
OR54611-5G
APOSOR54611-5GEA
183
EUR
APOSOR54611-5G
3-Acetylthiazolidine-2-thione
3-Acetylthiazolidine-2-thione
Formel:
C₅H₇NOS₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
76397-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...