Fmoc-(S)-nipecotic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-Nip-OH, (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-3-carboxylic acid

OR54626-5G
APOSOR54626-5GEA 159 EUR
APOSOR54626-5G
Fmoc-(S)-nipecotic acid
Fmoc-(S)-nipecotic acid
Formel: C₂₁H₂₁NO₄
Molecular Weight: 351.41 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04112682
CAS-Nummer: 193693-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR