5-Sulfoisophthalsäure Natriumsalz 98+%
Lieferant: Apollo Scientific
|
Achtung
|
OR54628-25G
OR54628-100G
APOSOR54628-25GEA
49.5
EUR
APOSOR54628-25G
APOSOR54628-100G
5-Sulfoisophthalsäure Natriumsalz 98+%
5-Sulfoisophthalsäure Natriumsalz
Formel:
C₈H₆O₇S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007495 CAS-Nummer: 6362-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...