Fmoc-D-Trp(Boc)-OH

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Boc-N-Fmoc-D-tryptophan

OR54631-1G
APOSOR54631-1GEA 21.9 EUR
APOSOR54631-1G
Fmoc-D-Trp(Boc)-OH
Fmoc-D-Trp(Boc)-OH
Formel: C₃₁H₃₀N₂O₆
Molecular Weight: 526.589 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00153367
CAS-Nummer: 163619-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR