7-Bromo-4-methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine

Lieferant: Apollo Scientific
Achtung

OR54640-1G
APOSOR54640-1GEA 244 EUR
APOSOR54640-1G
7-Bromo-4-methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
7-Bromo-4-methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
Formel: C₈H₉BrN₂O
Molecular Weight: 229,074 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025911
CAS-Nummer: 910037-14-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR