(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-Lys(Boc)-OH, Nα,Nepsilon-Di-Boc-L-lysine

OR54645-100G
APOSOR54645-100GEA 95.7 EUR
APOSOR54645-100G
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid 98%
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid
Formel: C₁₆H₃₀N₂O₆
Molecular Weight: 346,4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00038515
CAS-Nummer: 2483-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR