3-(Cyclopropyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

OR54650-1G OR54650-250MG
APOSOR54650-1GEA 97.3 EUR
APOSOR54650-1G APOSOR54650-250MG
3-(Cyclopropyl)benzyl alcohol
3-(Cyclopropyl)benzyl alcohol
Formel: C₁₀H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD06802678
CAS-Nummer: 893738-74-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR