3-Bromo-5-chloro-2-hydroxybenzonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
OR54658-1G
APOSOR54658-1GEA
282
EUR
APOSOR54658-1G
3-Bromo-5-chloro-2-hydroxybenzonitrile
3-Bromo-5-chloro-2-hydroxybenzonitrile
Formel:
C₇H₃BrClNO Molecular Weight: 232,462 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09705900 CAS-Nummer: 938117-34-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...