2-Hydroxy-4-cyclopropylbenzaldehyde

Lieferant: Apollo Scientific
Achtung

OR54662-250MG
APOSOR54662-250MGEA 256 EUR
APOSOR54662-250MG
2-Hydroxy-4-cyclopropylbenzaldehyde
2-Hydroxy-4-cyclopropylbenzaldehyde
Formel: C₁₀H₁₀O₂
Molecular Weight: 162.185 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31726536
CAS-Nummer: 2387695-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR