7-Amino-3,4-dihydro-2H-isoquinolin-1-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Amino-3,4-dihydroisoquinolin-1(2H)-one
OR54677-1G
OR54677-250MG
OR54677-5G
APOSOR54677-1GEA
224
EUR
APOSOR54677-1G
APOSOR54677-250MG
APOSOR54677-5G
7-Amino-3,4-dihydro-2H-isoquinolin-1-one 97%
7-Amino-3,4-dihydro-2H-isoquinolin-1-one
Formel:
C₉H₁₀N₂O Molecular Weight: 162.189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437645 CAS-Nummer: 66491-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...