7-Amino-3,4-dihydro-2H-isoquinolin-1-one 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Amino-3,4-dihydroisoquinolin-1(2H)-one

OR54677-1G OR54677-250MG OR54677-5G
APOSOR54677-1GEA 224 EUR
APOSOR54677-1G APOSOR54677-250MG APOSOR54677-5G
7-Amino-3,4-dihydro-2H-isoquinolin-1-one 97%
7-Amino-3,4-dihydro-2H-isoquinolin-1-one
Formel: C₉H₁₀N₂O
Molecular Weight: 162.189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437645
CAS-Nummer: 66491-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR