Tripropylsilan 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Tri-n-propylsilane
OR54684-25G
APOSOR54684-25GEA
174
EUR
APOSOR54684-25G
Tripropylsilan 99%
Tripropylsilan
Formel:
C₉H₂₂Si Molecular Weight: 158,36 g/mol Siedepunkt: 173 °C Dichte: 0,758 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009358 CAS-Nummer: 998-29-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...