2-Bromo-3-chlorobenzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR54687-100MG OR54687-1G OR54687-250MG
APOSOR54687-100MGEA 0 EUR
APOSOR54687-100MG APOSOR54687-1G APOSOR54687-250MG
2-Bromo-3-chlorobenzenesulphonyl chloride
2-Bromo-3-chlorobenzenesulphonyl chloride
Formel: C₆H₃BrCl₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD18390309
CAS-Nummer: 1261646-81-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR