3-(Methylsulfonyl)benzoesäure 97%

Lieferant: Apollo Scientific
Achtung

OR5476-100G OR5476-25G OR5476-5G
APOSOR5476-100GEA 386 EUR
APOSOR5476-100G APOSOR5476-25G APOSOR5476-5G
3-(Methylsulfonyl)benzoesäure 97%
3-(Methylsulphonyl)benzoic acid
Formel: C₈H₈O₄S
Schmelzpunkt: 230…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216488
CAS-Nummer: 5345-27-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR