4-Brom-2,6-xylenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Brom-2-hydroxy-3-xylol, 4-Brom-2,6-dimethylphenol
OR5479-10G
APOSOR5479-10GEA
21.9
EUR
APOSOR5479-10G
4-Brom-2,6-xylenol 98%
4-Brom-2,6-xylenol
Formel:
C₈H₉BrO Molecular Weight: 201,06 g/mol Schmelzpunkt: 74…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002314 CAS-Nummer: 2374-05-2 EINECS: 219-153-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...