2-(Methoxymethyl)aniline 95%

Lieferant: Apollo Scientific
Achtung

OR54820-1G OR54820-250MG
APOSOR54820-1GEA 545 EUR
APOSOR54820-1G APOSOR54820-250MG
2-(Methoxymethyl)aniline 95%
2-(Methoxymethyl)aniline
Formel: C₈H₁₁NO
Molecular Weight: 137.179 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09811778
CAS-Nummer: 62723-78-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR