2-Brom-3-chlortoluol

Lieferant: Apollo Scientific

Synonyme: 1-Brom-2-chlor-6-methylbenzol

OR5484-1G
APOSOR5484-1GEA 37.9 EUR
APOSOR5484-1G
2-Brom-3-chlortoluol
2-Brom-3-chlortoluol
Formel: C₇H₆BrCl
Molecular Weight: 205,48 g/mol
MDL: MFCD00060645
CAS-Nummer: 69190-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR