2'-Brom-4'-cyanacetanilid
Lieferant: Apollo Scientific
|
Achtung
|
OR5485-100MG
APOSOR5485-100MGEA
21.9
EUR
APOSOR5485-100MG
2'-Brom-4'-cyanacetanilid
2'-Brom-4'-cyanacetanilid
Formel:
C₉H₇BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094675 CAS-Nummer: 656834-86-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...