DL-2,3-Diaminopropionic acid hydrochloride 99%
Lieferant: Apollo Scientific
Synonyme:
2,3-Diaminopropanoic acid hydrochloride, 3-Amino-DL-alanine Hydrochloride, DL-2,3-Diaminopropionic acid monohydrochloride
OR5494-25G
APOSOR5494-25GEA
176
EUR
APOSOR5494-25G
DL-2,3-Diaminopropionic acid hydrochloride 99%
DL-2,3-Diaminopropionic acid hydrochloride
Formel:
C₃H₈N₂O₂ Molecular Weight: 140,57 g/mol Schmelzpunkt: 232 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012884 CAS-Nummer: 54897-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...