2-Brom-4-nitroanisol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-methoxy-4-nitrobenzene

OR5498-25G
APOSOR5498-25GEA 26.1 EUR
APOSOR5498-25G
2-Brom-4-nitroanisol
2-Brom-4-nitroanisol
Formel: C₇H₆BrNO₃
Molecular Weight: 232,03 g/mol
Schmelzpunkt: 103…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041242
CAS-Nummer: 5197-28-4
EINECS: 225-983-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR