2-Amino-2-(3,4-dichlorophenyl)ethanol 97%

Lieferant: Apollo Scientific

OR55015-250MG
APOSOR55015-250MGEA 241 EUR
APOSOR55015-250MG
2-Amino-2-(3,4-dichlorophenyl)ethanol 97%
2-Amino-2-(3,4-dichlorophenyl)ethanol
Formel: C₈H₉Cl₂NO
Molecular Weight: 206,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827189
CAS-Nummer: 188586-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR