2-Chloro-3-methylaniline 97%

Lieferant: Apollo Scientific
Achtung

OR55072-5G
APOSOR55072-5GEA 101 EUR
APOSOR55072-5G
2-Chloro-3-methylaniline 97%
2-Chloro-3-methylaniline
Formel: C₇H₈ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD11113332
CAS-Nummer: 29027-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR