(4-Methylquinazolin-2-yl)methanol 90%

Lieferant: Apollo Scientific
Achtung

OR55080-10MG
APOSOR55080-10MGEA 302 EUR
APOSOR55080-10MG
(4-Methylquinazolin-2-yl)methanol 90%
(4-Methylquinazolin-2-yl)methanol
Formel: C₁₀H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD18448900
CAS-Nummer: 13535-91-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR