4,6-Dichloro-2-methylpyrimidine-5-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR55083-5G
APOSOR55083-5GEA 88.6 EUR
APOSOR55083-5G
4,6-Dichloro-2-methylpyrimidine-5-carbaldehyde
4,6-Dichloro-2-methylpyrimidine-5-carbaldehyde
Formel: C₆H₄Cl₂N₂O
Molecular Weight: 191.015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909782
CAS-Nummer: 14160-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR