2-(2-Bromothiazol-4-yl)acetic acid

Lieferant: Apollo Scientific

Synonyme: 2-(2-bromo-1,3-thiazol-4-yl)acetic acid, (2-Bromo-thiazol-4-yl)-acetic acid

OR55097-100MG
APOSOR55097-100MGEA 288 EUR
APOSOR55097-100MG
2-(2-Bromothiazol-4-yl)acetic acid
2-(2-Bromothiazol-4-yl)acetic acid
Formel: C₅H₄BrNO₂S
Molecular Weight: 222,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18410768
CAS-Nummer: 62557-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR