Eicosanedioic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR55106-5G
OR55106-10G
APOSOR55106-5GEA
0
EUR
APOSOR55106-5G
APOSOR55106-10G
Eicosanedioic acid
Eicosanedioic acid
Formel:
C₂₀H₃₈O₄ Siedepunkt: 234 °C Schmelzpunkt: 127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059627 CAS-Nummer: 2424-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...