2-Azaspiro[3.4]octane hemioxalate

Lieferant: Apollo Scientific
Achtung

OR55128-1G
APOSOR55128-1GEA 989 EUR
APOSOR55128-1G
2-Azaspiro[3.4]octane hemioxalate
2-Azaspiro[3.4]octane hemioxalate
Formel: 2C₇H₁₃N·C₂H₂O₄
Molecular Weight: 312,405 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27988100
CAS-Nummer: 1523617-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR