5-Methoxyquinolin-2(1H)-one 95+%

Lieferant: Apollo Scientific
Achtung

OR55141-1G
APOSOR55141-1GEA 213 EUR
APOSOR55141-1G
5-Methoxyquinolin-2(1H)-one 95+%
5-Methoxyquinolin-2(1H)-one
Formel: C₁₀H₉NO₂
Molecular Weight: 175.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11877926
CAS-Nummer: 160893-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR