N-Boc-6-Bromohexylamine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
tert-Butyl (6-bromohexyl)carbamate, Boc-NH-C6-Br
OR55147-1G
APOSOR55147-1GEA
29
EUR
APOSOR55147-1G
N-Boc-6-Bromohexylamine
N-Boc-6-Bromohexylamine
Building block for preparing analogs of biologically active compounds
Formel:
C₁₁H₂₂BrNO₂ Molecular Weight: 280,202 g/mol Dichte: 1,192 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06201019 CAS-Nummer: 142356-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...