tert-Butyl 2-azaspiro[3.3]hept-6-ylcarbamate hemioxalate

Lieferant: Apollo Scientific
Achtung

OR55148-1G
APOSOR55148-1GEA 655 EUR
APOSOR55148-1G
tert-Butyl 2-azaspiro[3.3]hept-6-ylcarbamate hemioxalate
tert-Butyl 2-azaspiro[3.3]hept-6-ylcarbamate hemioxalate
Formel: 2C₁₁H₂₀N₂O₂·C₂H₂O₄
Molecular Weight: 514,612 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22421679
CAS-Nummer: 1523572-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR