3-(tert-Butoxycarbonylamino)-1-propanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Boc-amino)-1-propanol, N-(3-Hydroxypropyl)carbamic Acid tert-Butyl Ester, Boc-β-Ala-ol, 3-(BOC-amino)-1-propanol, tert-Butyl N-(3-Hydroxypropyl)carbamate
OR55160-100G
OR55160-25G
APOSOR55160-100GEA
212
EUR
APOSOR55160-100G
APOSOR55160-25G
3-(tert-Butoxycarbonylamino)-1-propanol 95%
3-(tert-Butoxycarbonylamino)-1-propanol
Formel:
C₈H₁₇NO₃ Molecular Weight: 175.23 g/mol Dichte: 1.025 g/cm³ (25 °C) Flammpunkt: 109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191883 CAS-Nummer: 58885-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...