22-(tert-Butoxy)-22-oxodocosanoic acid

Lieferant: Apollo Scientific
Achtung

OR55180-250MG
APOSOR55180-250MGEA 195 EUR
APOSOR55180-250MG
22-(tert-Butoxy)-22-oxodocosanoic acid
22-(tert-Butoxy)-22-oxodocosanoic acid
Formel: C₂₆H₅₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD30830215
CAS-Nummer: 1642333-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR