4-Chloro-2-(methylthio)-5-pyrimidinecarbonyl chloride 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-(methylthio)pyrimidine-5-carbonyl chloride

OR55187-5G
APOSOR55187-5GEA 190 EUR
APOSOR55187-5G
4-Chloro-2-(methylthio)-5-pyrimidinecarbonyl chloride 95%
4-Chloro-2-(methylthio)-5-pyrimidinecarbonyl chloride
Formel: C₆H₄Cl₂N₂OS
Molecular Weight: 223.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12545984
CAS-Nummer: 55084-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR