Boc-L-β-homovaline 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Boc-L-β-leucine, Boc-β-HoVal-OH, Boc-β-HomoVal-OH, (R)-3-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid

OR55192-5G
APOSOR55192-5GEA 144 EUR
APOSOR55192-5G
Boc-L-β-homovaline 98%
Boc-L-beta-homovaline
Formel: C₁₁H₂₁NO₄
Molecular Weight: 231.292 g/mol
Schmelzpunkt: 115…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01076232
CAS-Nummer: 183990-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR