Boc-D-trans-Hyp-OMe

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,4S)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate

OR55193-25G OR55193-5G
APOSOR55193-25GEA 192 EUR
APOSOR55193-25G APOSOR55193-5G
Boc-D-trans-Hyp-OMe
Boc-D-trans-Hyp-OMe
Formel: C₁₁H₁₉NO₅
Lagertemperatur: Kühlschrank
MDL: MFCD09954932
CAS-Nummer: 135042-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR