trans-4-Hydroxyzimtsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR55198-100G
OR55198-25G
OR55198-5G
APOSOR55198-100GEA
0
EUR
APOSOR55198-100G
APOSOR55198-25G
APOSOR55198-5G
trans-4-Hydroxyzimtsäure
trans-4-Hydroxyzimtsäure
Formel:
C₉H₈O₃ Schmelzpunkt: 214 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004399 CAS-Nummer: 501-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...